Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antifoam A, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
Description
Spectrum™ Chemical Antifoam A is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 100 mL |
| Packaging | Poly Bottle |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction