Antimony Broken rod, 12.7mm (0.50 in.) dia x various lengths, 99.8% (Metals basis), Alfa Aesar™

Form: Broken pieces from cast rod

Manufacturer: Alfa Aesar 4504736

Quantity 500g

 View more versions of this product

Catalog No. AA4504736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-36-0
    Physical Form Broken Rod, 12.7mm (0.50 in.) dia X various lengths
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00134030
    Chemical Name or Material Antimony

    Specifications

    Physical Form Broken Rod, 12.7mm (0.50 in.) dia X various lengths
    Quantity 500g
    Color Silver-gray
    Odor Odorless
    Assay Percent Range 99.8% (Metals basis)
    Appearance Silver Gray rod
    MDL Number MFCD00134030
    CAS 7440-36-0
    Chemical Name or Material Antimony

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit