Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antimony(III) acetate, 97%
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
Description
Antimony(III) acetate is a catalyst used in the production of synthetic fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 100 g |
UN Number | UN1549 |
Sensitivity | Moisture sensitive |
Solubility Information | Decomposes in water |
Formula Weight | 298.88 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | Antimony (III) acetate |
Safety and Handling
Hazard Category | H302+H332 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S. |
EINECSNumber | 230-043-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction