Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) acetate, 97%

Catalog No. p-7080374
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg

CAS: 6923-52-0 | C6H9O6Sb | 298.892 g/mol

Antimony(III) acetate is a catalyst used in the production of synthetic fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6923-52-0
Molecular Formula C6H9O6Sb
Molecular Weight (g/mol) 298.892
MDL Number MFCD00014974
InChI Key JVLRYPRBKSMEBF-UHFFFAOYSA-K
Synonym antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonity
PubChem CID 23354
IUPAC Name antimony(3+);triacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3]

Specifications

Quantity 10 g
UN Number UN1549
Sensitivity Moisture sensitive
Solubility Information Decomposes in water
Formula Weight 298.88
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Antimony (III) acetate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 230-043-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.