Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(Iii) Chloride, puriss. p.a.ACS Reagent, ≥99.0% (RT), Solstice

Catalog No. 60044657 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-657 100 g Poly Bottle
60-044-658 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-657 Supplier Solstice Supplier No. 10774100G

Puriss. p.a., ACS reagent,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 283°C
Melting Point 73.4°C
Color White
Quantity 100 g
Linear Formula SbCl3
UN Number UN1733
Formula Weight 228.12g/mol
Percent Purity ≥99.0% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Appearance Crystals/Powder
Vapor Pressure 1mmHg (49°C)
Density 3.140g/cu.cm; Vapor Density: 7.9 (vs air)
Chemical Name or Material Antimony(Iii) Chloride
Show More Show Less
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P501
Precautionary Statement H314-H411
DOTInformation 8/PGII
EINECSNumber 233-047-2
RTECSNumber CC4900000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.