Learn More
Antimony(III) oxide, 99%
Description
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

Specifications
Specifications
Boiling Point | 1550°C (sublimation) |
Melting Point | 656°C |
Quantity | 2500 g |
Merck Index | 14,711 |
Solubility Information | Insoluble in water. |
Formula Weight | 291.5 |
Percent Purity | 99% |
Odor | Odorless |
Density | 5.67 |
Chemical Name or Material | Antimony (III) oxide |
Safety and Handling
Hazard Category | H351 |
Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 215-175-0 |
RTECSNumber | CC5650000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.