Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) oxide, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
5kg
1309-64-4
O3Sb2
291.42
MFCD00011214
Antimony trioxide
1309-64-4
O3Sb2
291.42
MFCD00011214
Antimony trioxide
Assay Percent Range 99% min. (Sb2O3)
Linear Formula Sb2O3
Merck Index 15,699
Solubility Information Solubility in water: 2.7mg/l (20°C). Other solubilities: slightly soluble in hot water,soluble in conc. hcl,acetic acid,sulfuric acids,,solutions of alkali hydroxides
Formula Weight 291.42
Percent Purity 99+%
Packaging Plastic Bottle
Boiling Point 1550°C
Melting Point 656°C
Quantity 100g
Chemical Name or Material Antimony(III) oxide
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 215-175-