Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antimony(III) Oxide, Extra Pure, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
2.5 kg
50 kg
Packaging:
Fibre carton
Plastic bottle
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Formula Weight | 291.49 |
| Grade | Extra Pure |
| Odor | Odorless |
| Packaging | Plastic bottle |
| Appearance | White solid |
| Vapor Pressure | 13.3 hPa at 660°C |
| Chemical Name or Material | Antimony(III) oxide |
Safety and Handling
| Hazard Category | H351: Suspected of causing cancer. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction