Alfa Aesar™ Antimony(III) selenide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1313006

Quantity 5g

 View more versions of this product

Catalog No. AA1313006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1315-05-5
    Formula Weight 480.38
    MDL Number MFCD00016319
    UN Number UN3077
    Merck Index 14,712
    Chemical Name or Material Antimony(III) selenide

    Specifications

    Formula Weight 480.38
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    TSCA TSCA
    UN Number UN3077
    Melting Point 611°C
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Particle Size 3 to 12mm
    MDL Number MFCD00016319
    Merck Index 14,712
    Quantity 5g
    Solubility Information Very slightly soluble in water
    Physical Form Pieces
    Sensitivity Moisture Sensitive
    CAS 1315-05-5
    Chemical Name or Material Antimony(III) selenide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H330-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit