Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(III) sulfide, 99.999% (metals basis)

Catalog No. p-7076483
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Plastic bottle

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

Antimony(III) sulfide is manufactured for use in safety matches, military ammunition, explosives and fireworks. Also it is used in the production of ruby-colored glass and in plastics as a flame retardant. It is a semiconductor with a direct band gap of 1.8-2.5 eV. With suitable doping p and n type materials can be produced.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Packaging Glass bottle
Quantity 10 g
UN Number UN1549
Solubility Information Insoluble in water.
Formula Weight 339.68
Percent Purity 99.999%
Physical Form Pieces
Chemical Name or Material Antimony(III) sulfide
Hazard Category H302-H312-H315-H319-H335-H500
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 215-713-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.