Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony pentafluoride, 99+%

Catalog No. AC174431000
Click to view available options
Quantity:
100 g

CAS: 7783-70-2 | F5H8Sb | 224.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-70-2
Molecular Formula F5H8Sb
Molecular Weight (g/mol) 224.82
MDL Number MFCD00011219
InChI Key VBKPQOIVRXDIEL-UHFFFAOYSA-N
Synonym antimony pentafluoride, antimony v fluoride, pentafluoroantimony, antimony v pentafluoride, antimony fluoride sbf5, hsdb 442, antimonypentafluoride, pentafluorostibine, pentafluorostiborane
IUPAC Name antimony pentafluoride
SMILES F.F.F.F.F.[SbH3]

Specifications

Boiling Point 149.4°C
Melting Point 7.0°C
Color White
Physical Form Shiny Flakes or Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula SbF5
Fieser 05,19; 06,23; 09,2
Merck Index 15, 687
Solubility Information Solubility in water: reacts with water. Other solubilities: forming hf and antimonyoxide, soluble in kf and liquid so2
Formula Weight 216.74
Percent Purity ≥99%
Packaging Plastic Bottle packed in tin can
Specific Gravity 2.99
Density 2.9900g/mL
Chemical Name or Material Antimony pentafluoride, 99.5+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 232-021-8
RTECSNumber CC5800000
TSCA TSCA
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.