Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%

Catalog No. aa4061914
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 28300-74-5 | C8H4K2O12Sb2·3H2O

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2·3H2O
MDL Number MFCD00148863
Synonym Potassium antimonyl tartrate

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99.0-103.0%
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 667.87 (613.82 Anhydrous)
Grade ACS
Density 2.6
Chemical Name or Material Antimony potassium tartrate trihydrate
Hazard Category H302+H332-H335-H373
Hazard Statement Avoid release to the environment. Refer to special instructions/Safety data sheets
Precautionary Statement P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 229-436-1
RTECSNumber CC6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.