Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony tin oxide, NanoArc™, 99.5%

Catalog No. p-7059588
Click to view available options
Quantity:
25 g
100 g
500 g

Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge.Antimony tin oxide is used in liquid crystal display, equipment configuration database and light emitting diode. It is used in the glass of train, car and plane, which used to protect against fog and frost. It is used to protect the electromagnetic wave in the radar shielding and protecting area. It is used in the field of optoelectronic display devices, transparent electrode, solar battery and liquid crystal display.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Moderately conductive particle
  • The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony tin oxide NanoTek™
Physical Form Powder
Quantity 25 g
MDL Number MFCD00799153
UN Number UN1549
Solubility Information Insoluble in water.
Percent Purity 99.5%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.