Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Antimony, Trichloride
SCI_ED

Catalog No. s25183 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Crystals

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name antimony(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Physical Form Solid
Quantity 100 g
Assay Percent Range 0.99
Linear Formula SbCl3
Formula Weight 228.11
Packaging Crystals
Chemical Name or Material Antimony, Trichloride
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.