Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony trichloride, 99.5%, ACROS Organics™

Click to view available options
Quantity:
500g
2kg
10kg
Packaging:
Glass bottle
Plastic Bottle
10025-91-9
Cl3Sb
228.11
MFCD00011212
FAPDDOBMIUGHIN-UHFFFAOYSA-K
antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony
24814
CHEBI:74856
trichlorostibane
Cl[Sb](Cl)Cl
10025-91-9
Cl3Sb
228.11
MFCD00011212
FAPDDOBMIUGHIN-UHFFFAOYSA-K
antimony trichloride,trichlorostibine,antimony chloride,antimony iii chloride,stibine, trichloro,antimontrichlorid,antimonous chloride,butter of antimony,antimony butter,caustic antimony
24814
CHEBI:74856
trichlorostibane
Cl[Sb](Cl)Cl
Assay Percent Range 99.4% min. (Iodimetry)
Linear Formula SbCl3
Fieser 08,19
Merck Index 15, 696
Arsenic (As) 100ppm max.
Solubility Information Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,,soluble in chloroform (+/- 22%),soluble in alcohol,benzene,carbon disulfide,,acetone,tartaric acid,ccl4,insoluble in pyridine,quinoline,organic bases
Formula Weight 228.11
Physical Form Liquid
Percent Purity 99.50%
Insoluble Matter 0.01% max. (in HCl)
Iron (Fe) 10ppm max.
Packaging Glass bottle
Boiling Point 223°C
Color Colorless
Melting Point 73°C
Quantity 500g
Chemical Name or Material Antimony trichloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-047-2