Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony trichloride, ACS reagent

Catalog No. AC401370025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401370025 2.5 kg Glass Bottle
AC401370050 5 g Glass Bottle
AC401371000 100 g Glass Bottle
AC401375000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC401370025 Supplier Thermo Scientific Chemicals Supplier No. 401370025
Add to Cart

CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.0°C
Melting Point 73.0°C
Color White
Physical Form Powder, Crystals and/or Chunks
Quantity 2.5 kg
Linear Formula SbCl3
UN Number 1733
Fieser 08,19
Merck Index 15,696
Solubility Information Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,, soluble in chloroform (+/- 22%), soluble in alcohol, benzene, carbon disulfide,, acetone, tartaric acid, ccl4, insoluble in pyridine, quinoline, organic bases
IUPAC Name trichlorostibane
Formula Weight 228.11
Percent Purity ≥99.0% (Iodimetry)
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Antimony trichloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-047-2
RTECSNumber CC4900000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.