Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony trifluoride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Plastic bottle
7783-56-4
F3Sb
178.75
MFCD00011218
GUNJVIDCYZYFGV-UHFFFAOYSA-K
antimony trifluoride,trifluorostibine,antimony fluoride,antimony iii fluoride,stibine, trifluoro,trifluoroantimony,antimoine fluorure,antimonous fluoride,antimoine fluorure french,hsdb 438
24554
trifluorostibane
F[Sb](F)F
7783-56-4
F3Sb
178.75
MFCD00011218
GUNJVIDCYZYFGV-UHFFFAOYSA-K
antimony trifluoride,trifluorostibine,antimony fluoride,antimony iii fluoride,stibine, trifluoro,trifluoroantimony,antimoine fluorure,antimonous fluoride,antimoine fluorure french,hsdb 438
24554
trifluorostibane
F[Sb](F)F
Assay Percent Range 97.5% min. (Iodimetry)
Linear Formula SbF3
Fieser 05,526; 06,553
Merck Index 15, 697
Solubility Information Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water,insoluble in nh3
Formula Weight 178.75
Physical Form Crystalline Powder
Percent Purity 98%
Packaging Plastic bottle
Solubility Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water, insoluble in nh3
Sublimation Point 376 °C
Boiling Point 376.0°C
Color White to Gray
Melting Point 292.0°C
Quantity 100g
Chemical Name or Material Antimony trifluoride, 98%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 232-009-2

RTECSNumber : CC5150000

TSCA : TSCA