Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antimony trifluoride, 98%
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 376.0°C |
| Melting Point | 292.0°C |
| Color | White to Gray |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 97.5% min. (Iodimetry) |
| Linear Formula | SbF3 |
| Fieser | 05,526; 06,553 |
| Merck Index | 15, 697 |
| Solubility Information | Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water, insoluble in nh3 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic in contact with skin. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Toxic if swallowed. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: |
| EINECSNumber | 232-009-2 |
| RTECSNumber | CC5150000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction