Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC434528000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Antimony(V)-chloride
Color Yellow
CAS 7647-18-9, 75-09-2
Physical Form Solution
Quantity 800 mL
Molecular Formula Cl5Sb
Linear Formula SbCl5
MDL Number MFCD00011213
Merck Index 15,686
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl
IUPAC Name pentachloro-$l^{5}-stibane
Molecular Weight (g/mol) 299.02
PubChem CID 24294
Formula Weight 299.01
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.442
Density 1.4420g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.