Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony(V) oxide, Puratronic™, 99.998% (metals basis)

Catalog No. p-4407250
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1314-60-9 | O5Sb2 | 323.515 g/mol

It is used as flame retardants in acrylonitrile-butadiene-styrene (ABS) polymer, plastics, rubbers, paper and textiles. It finds use as a flocculant in the production of titanium dioxide. It also finds use in ion-exchange resin. It is also employed as catalyst in certain polymerization and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-60-9
Molecular Formula O5Sb2
Molecular Weight (g/mol) 323.515
MDL Number MFCD00011216
InChI Key LJCFOYOSGPHIOO-UHFFFAOYSA-N
Synonym antimony pentoxide, diantimony pentoxide, antimonic oxide, antimony v oxide, diantimony pentaoxide, stibic anhydride, antimony pentaoxide, apox s, antimony oxide sb2o5, nyacol ago 40
PubChem CID 14813
IUPAC Name (dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
SMILES O=[Sb](=O)O[Sb](=O)=O

Specifications

Melting Point 380°C (decomposition)
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3
Formula Weight 323.5
Odor Odorless
Appearance Yellowish powder
Density 3.78
Chemical Name or Material Antimony(V) oxide
Show More Show Less
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 215-237-7
RTECSNumber CC6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.