Learn More
Antimony(V) oxide, Puratronic™, 99.998% (metals basis)
Description
It is used as flame retardants in acrylonitrile-butadiene-styrene (ABS) polymer, plastics, rubbers, paper and textiles. It finds use as a flocculant in the production of titanium dioxide. It also finds use in ion-exchange resin. It is also employed as catalyst in certain polymerization and oxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 380°C (decomposition) |
Color | Yellow |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3 |
Formula Weight | 323.5 |
Odor | Odorless |
Appearance | Yellowish powder |
Density | 3.78 g/cm3 |
Show More |
Safety and Handling
Hazard Category | H302+H332 |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: III |
EINECSNumber | 215-237-7 |
RTECSNumber | CC6300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.