Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Antimycin A

Click to view available options
Quantity:
5mg
10mg
1397-94-0
C28H40N2O9
548.633
MFCD01779723
UIFFUZWRFRDZJC-RBVQMQRASA-N
16218979
[(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate
CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C
1397-94-0
C28H40N2O9
548.633
MFCD01779723
UIFFUZWRFRDZJC-RBVQMQRASA-N
16218979
[(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate
CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C
Applications
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

Solubility
Soluble in 100% ethanol. Insoluble in water.

Notes
Isolated from Streptomyces sp.
UN Number UN3462
Merck Index 14,714
Formula Weight 548.63
Physical Form Powder
Color White to Yellow
Melting Point 149°C to 150°C
Quantity 10mg
Sensitivity Light Sensitive
Chemical Name or Material Antimycin A

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.

RTECSNumber : CD0350000

Recommended Storage : −30° to −10°C