Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, 98%

Catalog No. p-7081212
Click to view available options
Quantity:
100 g
500 g

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

Melting Point 111°C to 114°C
Quantity 100 g
Beilstein 157775
Merck Index 14,716
Formula Weight 188.23
Percent Purity 98%
Chemical Name or Material Antipyrine
Hazard Category H302-H315-H317-H319-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 200-486-6
RTECSNumber CD2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.