Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antipyrine, 99%

Catalog No. AC104975000
Click to view available options
Quantity:
100 g
500 g

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

Melting Point 109°C to 113°C
Color White
Boiling Point 319°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodimetry)
Packaging Plastic bottle
Quantity 500 g
Beilstein 24,27
Merck Index 15,703
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and chloroform,slightly soluble in ether
Formula Weight 188.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Antipyrine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
EINECSNumber 200-486-6
RTECSNumber CD2450000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.