Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antipyrine, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Pharmaceutical (small molecule)

Specifications
Specifications
Packaging | Poly Bottle |
CAS | 60-80-0 |
Physical Form | Neat |
Linear Formula | C11H12N2 O |
MDL Number | MFCD00003146 |
Melting Point | 109°C to 111°C |
Synonym | 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one; Phenazone |
Type | Analytical Standard |
Quantity | 100 g |
Formula Weight | 188.23 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction