Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Antizyme inhibitor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115481AP150UL

Description
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.Specifications
Antizyme inhibitor 1 | |
Polyclonal | |
Unconjugated | |
AZIN1 | |
Antizyme inhibitor 1, AZI, AZIN1, OAZI, OAZIN, ODC1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51582, 54375, 58961 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14977, O35484, Q63764 | |
AZIN1 | |
Antizyme inhibitor 1 Fusion Protein Ag2082 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction