Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Antp Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569514
Description
This target displays homology in the following species: Fruit fly:100%;.
Antp gene ontology annotations related to this gene include multicellular organism development.
Specifications
Antp | |
Polyclonal | |
Unconjugated | |
Antp | |
3.4; Ant; ANTC; ANT-C; antenapedia; antennapaedia; antennapedia; antennapedia P1; antennapedia P2; antennapedia protein; antp; ANT-P; Antp P1; Antp P2; AntP1; Antp-PD; Antp-PE; Antp-PF; Antp-PG; Antp-PH; Antp-PI; Antp-PJ; Antp-PK; Antp-PL; Antp-PM; Antp-PN; Antp-PO; atennapedia; Aus; BG:DS07700.1; CG1028; CG1028-PD; CG1028-PE; CG1028-PF; CG1028-PG; CG1028-PH; CG1028-PI; CG1028-PJ; CG1028-PK; CG1028-PL; CG1028-PM; CG1028-PN; CG1028-PO; Dm-Antennapedia; DmAntp; DMANTPE1; Dmel\CG1028; Dmel_CG1028; DRO15DC96Z; Homeotic protein antennapedia; Hu; humeral; l(3)84Ba; LD33666p; nasobemia; Ns; Scx; Sex comb extra; Sex combs extra | |
Rabbit | |
Affinity chromatography | |
RUO | |
40835 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P02833 | |
Antp | |
synthetic peptide corresponding to a region of fruit fly Antp (aa 151-200). | |
100 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction