Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANXA1 Mouse anti-Human, Clone: OTI3A8, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500967

Description
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.Specifications
ANXA1 | |
Monoclonal | |
0.9 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P04083 | |
ANXA1 | |
Full length human recombit protein of human ANXA1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3A8 | |
Unconjugated | |
ANXA1 | |
ANX1, LPC1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
301 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction