Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANXA1 Mouse anti-Human, Unconjugated, Clone: ANXA1/1672, Abnova™

Catalog No. 89337713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-713 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-713 Supplier Abnova Corporation Supplier No. MAB21220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ANXA1 Monoclonal antibody specifically detects ANXA1 in Human samples. It is validated for ELISA.

Specifications

Antigen ANXA1
Applications ELISA
Classification Monoclonal
Clone ANXA1/1672
Conjugate Unconjugated
Description annexin A1
Gene ANXA1
Gene Alias ANX1, LPC1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human ANXA1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 301
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.