Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ANXA2 Mouse anti-Bovine, Human, Clone: Z014, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13988
Description
Mouse monoclonal antibody raised against bovine ANXA2.
- High specificity
- High reactivity
Specifications
ANXA2 | |
Monoclonal | |
Unconjugated | |
ELISA (0.1-1.0 ug/mL) Immunohistochemistry (5-10 ug/mL) Western Blot (0.5-1.0 ug/mL) The optimal working dilution should be determined by the end user. | |
P04272 | |
Mouse | |
Affinity purification | |
RUO | |
282689 | |
Bovine, Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
Z014 | |
Mouse monoclonal antibody raised against bovine ANXA2. | |
In proprietary buffer (0.09% sodium azide). | |
ANXA2 | |
Native purified ANXA2 from bovine lung. | |
100 μg | |
Primary | |
This antibody is specific for ANXA2 (also known as: calpactin I, chromobindin 8, p36, PAP-IV, and Protein I). It does not cross-react with other Annexin proteins. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction