Learn More
ANXA3 Mouse anti-Human, Clone: OTI2F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502122

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Specifications
ANXA3 | |
Monoclonal | |
Unconjugated | |
ANXA3 | |
ANX3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12429 | |
ANXA3 | |
Full length human recombit protein of human ANXA3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.