Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANXA5 Mouse anti-Human, Unconjugated, Clone: 1F4-1A5, Abnova™

Catalog No. 89335016 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-016 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-016 Supplier Abnova Corporation Supplier No. H00000308M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ANXA5 Monoclonal antibody specifically detects ANXA5 in Human samples. It is validated for WB-Ce, IHC-P, IF, S-ELISA, ELISA, WB-Re, WB-Tr, IP.

Specifications

Antigen ANXA5
Applications Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 1F4-1A5
Conjugate Unconjugated
Description annexin A5
Gene ANXA5
Gene Accession No. BC001429
Gene Alias ANX5, ENX2, PP4
Host Species Mouse
Immunogen ANXA5 (AAH01429, 1 a.a. ∼ 320 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 308
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.