Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANXA8L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571353 Supplier Invitrogen™ Supplier No. PA571353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANXA8L1 Polyclonal Antibody for Western Blot

This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANXA8L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANXA8L1
Gene Accession No. Q5VT79
Gene Alias Annexin A8; annexin A8L2; annexin A8-like 1; annexin A8-like protein 1; annexin A8-like protein 2; annexin VIII; Annexin-8; ANX8; ANXA8; ANXA8L1; ANXA8L2; bA145E20.2; VAC-beta; Vascular anticoagulant-beta
Gene Symbols ANXA8L1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ANXA8L1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728113
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.