Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANXA8L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596743 Supplier Invitrogen™ Supplier No. PA596743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAWWKAWIEQ EGVTVKSSSH FNPDPDAETL YKAMKGIGVG SQLLSHQAAA FAFPSSALTS VSPWGQQGHL CCNPAGTNEQ AIIDVLTKRS NTQRQQIAKS FKAQFGKDLT ETLKSELSGK FERLIVALMY PPYRYEAKEL HDAMKGSRDD VSSFVDPALA LQDAQDLYAA GEKIRGTDEM KFITILCTRS ATHLLRVKCT QNLHSYFAER LYYAMKGAGT RDGTLIRNIV SRSEIDLNLI KCHFKKMYGK TLSSMIMEDT SGDYKNALLS LVGSDP; Positive Samples: A-549, HeLa, Mouse lung, Mouse ovary.

This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANXA8L2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANXA8L1
Gene Accession No. Q5VT79
Gene Alias AI466995; annexin 8; annexin A8; annexin A8L2; annexin A8-like 1; annexin A8-like protein 1; annexin A8-like protein 2; annexin VIII; Annexin-8; Anx8; ANXA8; ANXA8L1; ANXA8L2; bA145E20.2; LOC100052045; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPTF-associated factor 65 gamma; STAGA complex 65 subunit gamma; suppressor of Ty 7-like; SUPT7L; Unknown (protein for MGC:137593); VAC-beta; Vascular anticoagulant-beta
Gene Symbols ANXA8L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human ANXA8L2 (NP_0012658531).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728113
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.