Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ANXA8L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568746
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.
This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
Specifications
| ANXA8L2 | |
| Polyclonal | |
| Unconjugated | |
| ANXA8L1 | |
| AI466995; annexin 8; annexin A8; annexin A8L2; annexin A8-like 1; annexin A8-like protein 1; annexin A8-like protein 2; annexin VIII; Annexin-8; Anx8; ANXA8; ANXA8L1; ANXA8L2; bA145E20.2; LOC100052045; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPTF-associated factor 65 gamma; STAGA complex 65 subunit gamma; suppressor of Ty 7-like; SUPT7L; Unknown (protein for MGC:137593); VAC-beta; Vascular anticoagulant-beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728113 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5VT79 | |
| ANXA8L1 | |
| synthetic peptide directed towards the N terminal region of human ANXA8L2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction