Learn More
Stoelting™ ANY-box Behavior Test System
SDP
Stoelting™ Behavior Test System is a completely automated system designed to make behavioral tracking convenient and affordable without compromising features.
$38600.00 - $46000.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 2
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
Catalog Number Mfr. No. Model Size Price Quantity & Availability  
10-000-378
SDP
View Documents
Stoelting™
68500
4-behavior test system 100 cm
Each for $38,600.00
Only null left
Add to Cart
10-000-372
SDP
View Documents
Stoelting™
65500
5-behavior test system 40 cm
Each for $46,000.00
Only null left
Add to Cart
Description
Description
  • At the heart of ANY-box is the ANY-maze software, coupled to Stoelting's ANY-maze interface (AMi-2) device.
  • This combination of core components allows you to automate as many as four ANY-boxes per digial interface for simultaneous testing.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.