Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AOAH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587845 Supplier Invitrogen™ Supplier No. PA587845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFLDSTVGIK EKSIYLRLWK RNHCNHRDYQ NISRNGASSR NLKKFIESLS RNKVLDYPAI VIYAMIGNDV CSGKSDPVPA MTTPEKLYSN VMQTLKHLNS HLPNGSHVIL YGLPDGTFLW DNLHNRYHPL GQLNKDMTYA QLYSFLNCLQ VSPCHGWMSS NKTLRTLTSE RAEQLSNTLK KIAASEKFTN FNLFYMDFAF HEIIQEWQKR GGQPWQLIEP VDGFHPNEVA LLLLADHFWK KVQLQWPQIL GKENPFNPQI KQVFGDQGGH; Positive Samples: BxPC-3.

Removes the secondary (acyloxyacyl-linked) fatty acyl chains from the lipid A region of bacterial lipopolysaccharides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOAH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AOAH
Gene Accession No. P28039
Gene Alias 4930433E13Rik; Acyloxyacyl hydrolase; acyloxyacyl hydrolase (neutrophil); Acyloxyacyl hydrolase large subunit; Acyloxyacyl hydrolase small subunit; AOAH; RGD1560514
Gene Symbols AOAH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 306-575 of human AOAH (NP_0016281).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.