Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AOC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535855
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Bovine (80%).
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes and ammonia in the presence of copper and quinone cofactor. This gene shows high sequence similarity to copper amine oxidases from various species ranging from bacteria to mammals. The protein contains several conserved motifs including the active site of amine oxidases and the histidine residues that likely bind copper. It may be a critical modulator of signal transmission in retina, possibly by degrading the biogenic amines dopamine, histamine, and putrescine. This gene may be a candidate gene for hereditary ocular diseases. Alternate splicing results in multiple transcript variants. [provided by RefSeq].
Specifications
| AOC2 | |
| Polyclonal | |
| Unconjugated | |
| AOC2 | |
| Amine oxidase [copper-containing]; amine oxidase copper containing 2; amine oxidase, copper containing 2; amine oxidase, copper containing 2 (retina-specific); AOC2; DAO2; RAO; retina-specific amine oxidase; retina-specific copper amine oxidase; Semicarbazide-sensitive amine oxidase; SSAO | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.94 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O75106 | |
| AOC2 | |
| Recombinant protein encompassing a sequence within the center region of human AOC2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction