Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AOCS-Tintometer Colour Reference Standard 1.3R 8.8Y (5Â1⁄4), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
500 mL
Description
Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications
Specifications
CAS | 7647-01-0 |
Type | Color Reference Standard |
Quantity | 500 mL |
Grade | Certified Reference Material |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction