Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Evonik Corporation AOF L-Alanyl-L-Tyrosine Dihydrate (2H2O)
Click to view available options
Quantity:
20 kg
Description
- CAS #: 3061-88-9
- Assay (by HCI04 titration): 97.0 to 103.0%
- Bacterial endotoxins test: nmt. 10.0 EU/g
- Not to be used as API
- Animal Origin Free

Specifications
Specifications
CAS | 3061-88-9 |
Bacterial Endotoxins | nmt. 10.0 EU/g |
Assay Percent Range | 97.0 to 103.0% |
Quantity | 20 kg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction