Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559102 Supplier Invitrogen™ Supplier No. PA559102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYHALLKHLG TLAGSQIRNM ASLGGHIISR HPDSDLNPIL AVGNCTLNLL SKEGKRQIPL NEQFLSKCPN ADLKPQEILV SVNIPYSRKW E Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AOX1
Gene Accession No. Q06278
Gene Alias AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro
Gene Symbols AOX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AOX1. Recombinant protein control fragment (Product #RP-96097).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.