Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AOX1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598464 Supplier Invitrogen™ Supplier No. PA598464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AOX1A Polyclonal Antibody for Western Blot, ELISA

Catalyzes the cyanide-resistant oxidation of ubiquinol and the reduction of molecular oxygen to water, but does not translocate protons and consequently is not linked to oxidative phosphorylation. Increases respiration when the cytochrome respiratory pathway is restricted, or in response to low temperatures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOX1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias alternative oxidase 1A; AT3G22370; ATAOX1A; AtHSR3; HSR3; hyper-sensitivity-related 3
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana Ubiquinol oxidase 1a, mitochondrial protein (63-178AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.