Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AP 18
GREENER_CHOICE

Catalog No. 329610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-961-0 10 mg
32-965-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-961-0 Supplier Tocris Bioscience™ Supplier No. 3296/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reversible TRPA1 channel blocker

AP 18 is a reversible TRPA1 channel blocker (IC50 values are 3.1 and 4.5 μM at human and mouse TRPA1 respectively). Blocks cinnamaldehyde-induced but not capsaicin-induced nociception and reverses mechanical hyperalgesia in vivo. Also blocks TRPA1 pore dilation (IC50 = 10.3 μM for the inhibition of Yo-Pro uptake).

Chemical Identifiers

CAS 55224-94-7
Molecular Formula C11H12ClNO

Specifications

Quantity 10 mg
Formula Weight 209.67
Purity >98%
Molecular Weight (g/mol) 209.673
Chemical Name or Material AP 18
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.