Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AP 1867
GREENER_CHOICE

Catalog No. 620710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-071-0 10 mg
62-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-071-0 Supplier Tocris Bioscience™ Supplier No. 6207/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective binding ligand for FKBP12F36V

AP 1867 is a selective binding ligand for the single point mutant of FKBP12F36V (IC50 = 1.8 nM). Functionalized with a carboxylic acid group at the meta-position to enable onward chemistry. The position of the carboxylic acid group represents an 'exit vector' allowing modification without interfering with the compound's binding ability.

Specifications

Chemical Name or Material (1R)-1-[3-(Carboxymethoxy)phenyl]-3-(3,4-dimethoxyphenyl)propyl (2S)-1-[(2S)-1-oxo-2-(3,4,5-trimethoxyphenyl)butyl]-2-piperidinecarboxylate
CAS 195514-23-9
Quantity 10 mg
Target TAG Degradation Platforms
Molecular Formula C38H47NO11
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.