Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AP-2 beta/TFAP2B Antibody (6F7G1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | AP-2 beta/TFAP2B |
---|---|
Clone | 6F7G1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61858
![]() |
Novus Biologicals
NBP261858 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
AP-2 beta/TFAP2B Monoclonal specifically detects AP-2 beta/TFAP2B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
AP-2 beta/TFAP2B | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
activating enhancer binding protein 2 beta, Activating enhancer-binding protein 2-beta, AP-2B, AP2-B, AP2-beta, CHAR, MGC21381, transcription factor AP-2 beta (activating enhancer binding protein 2 beta), transcription factor AP-2 beta (activating enhancer-binding protein 2 beta), transcription factor AP-2-beta | |
TFAP2B | |
IgG2b | |
Protein G purified |
6F7G1 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Neuroscience | |
7021 | |
Purified recombinant fragment of human TFAP2B (AA: 84-193) expressed in E. Coli. | |
Primary | |
50.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title