Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AP 24534
GREENER_CHOICESDP

Catalog No. 427410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-741-0 10 mg
42-745-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-741-0 Supplier R&D Systems™ Supplier No. 4274/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent multi-kinase and pan-Bcr-Abl inhibitor

Potent multi-kinase and pan-Bcr-Abl inhibitor. Displays potent activity against cell lines expressing native Bcr-Abl or Bcr-AblT3151 (IC50 values are 0.37 and 2.0 nM respectively); also inhibits other Abl kinase domain mutants at nanomolar potencies. Exhibits inhibitory activity against PDGFRα, c-Src and c-Kit (IC50 values are 1.1, 5.4 and 12.5 nM respectively); potently inhibits FGFR and VEGFR family kinases. Orally active. Identified as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 943319-70-8
Molecular Formula C29H27F3N6O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 532.56
Purity >98%
Molecular Weight (g/mol) 532.571
Chemical Name or Material AP 24534
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.