Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Kits: Beer's Law and the Rate Law of Crystal Violet
GREENER_CHOICESCI_ED

Catalog No. S07320 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07320 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Crystal Violet, which is known to the State of California to cause cancer.
Catalog No. S07320 Supplier Innovating Science™ Supplier No. IS8111
Add to Cart
Add to Cart
This item is not returnable. View return policy
WARNING: This product can expose you to chemicals including Crystal Violet, which is known to the State of California to cause cancer.

Includes

Crystal violet 1% Aq. (3 x 500mL), Sodium hydroxide 0.2M (3 x 500mL), Deionized water (2 x 500mL), Teacher's guide, Student study guide copymasters

Requires

Beaker (50mL), cuvettes, pipet bulb, serological pipet (1mL), spectrophotometer or colorimeter with data collection system, stirring rod, stopwatch (if not using data collection software), volumetric flask (1000mL)

Determine the rate law for the reaction of crystal violet and sodium hydroxide.

Students can also prepare dilutions of stock crystal violet solutions to generate a Beer's law calibration curve. This lab will require students to integrate prior chemistry knowledge involving spectroscopy, Beer's law, solution dilution, calibration curves, and chemical kinetics.

  • Meets Big Idea 4, Investigation 11, Primary Learning Objective 4.2
  • Contains materials for 15 groups of students

Specifications

Certifications/Compliance Meets big idea 4, investigation 11, primary learning objective 4.2.
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Determine the rate law for the reaction of crystal violet and sodium hydroxide
Includes 1500ml 1% crystal violet, 1500ml 0.2m sodium hyroxide, 1000ml deionized water, teacher's guide and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 10 x 10 x 10 in.
Class Size 15 Lab Groups
Product Type Beer'S Law and the Rate Law of Crystal Violet Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.