Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Environmental Science: Net Primary Productivity of Aquatic Plants
SCI_ED

Catalog No. S000821 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Net Primary Productivity of Aquatic Plants
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000821 Net Primary Productivity of Aquatic Plants
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, which are known to the State of California to cause cancer.
Catalog No. S000821 Supplier Innovating Science™ Supplier No. IS9908
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, which are known to the State of California to cause cancer.

In this activity, simulate the photosynthesis and cellular respiration of an aquatic plant under different conditions to demonstrate when plants need energy for growth, respiration, reproduction and movement.

Living organisms require energy for growth, respiration, reproduction, and movement. Primary producers such as plants absorb energy from the sun and convert it to biomass via photosynthesis, which is then either used for cellular respiration or passed up the food chain.

  • In this activity, simulate the photosynthesis and cellular respiration of an aquatic plant under different conditions to demonstrate when plants need energy for each process.
  • This kit contains enough materials for 8 groups of students and comes with a Teacher's Manual and Student Study Guide Copymaster.
  • The activity covers AP Environmental Science primary learning objective ENG-1.A.
  • It will take a total of about 75 minutes over the course of 3 to 4 days to complete this activity (Day 1: 45 min, Day 3 or 4: 30 min).

Kit contains:

  • 2 Manganese Chloride Reagents, 25 mL
  • 2 Alkaline Iodide Reagents, 25 mL
  • 2 Sulfuric Acid, 50%, 25 mL
  • 2 Starch Indicator Solution, 15 mL
  • 2 Sodium Thiosulfate Titrant, 0.002 M, 750 mL
  • 24 Pipettes
  • 16 Sample Bottles with Caps

Needed but not supplied:

  • Bucket, 5 Gallon or Similar (for Class Water Sample)
  • Graduated Cylinders, 25 mL
  • Burettes, 50 mL
  • Burette Clamps/Stands
  • Funnels
  • Erlenmeyer Flasks, 125 mL
  • Elodea or Other Aquatic Plant - Can be collected locally
  • Scissors
  • Paper Towels
  • Markers
  • Aluminum Foil
  • Magnetic Stir Plate and Stir Bar (Optional)
  • Grow Light (Optional)

Specifications

Grade Level High School
Grade 9 to 12
Class Size 8 Lab Groups
Product Type Net Primary Productivity of Aquatic Plants
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.