Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Environmental Science: Physical and Chemical Properties of Soil
SCI_ED

Catalog No. S000824 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Properties of Soil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000824 Properties of Soil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable crystalline silica and Methanol, which are known to the State of California to cause cancer, birth defects, or other reproductive harm.
Catalog No. S000824 Supplier Innovating Science™ Supplier No. IS9902
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable crystalline silica and Methanol, which are known to the State of California to cause cancer, birth defects, or other reproductive harm.

Perform chemical and physical tests on local soil samples to explore different soil types and their characteristics due to their unique compositions.

Perform chemical and physical tests on local soil samples to explore different soil types and their characteristics due to their unique compositions.

  • This kit contains enough materials for 8 groups of students and comes with a Teacher's manual and Student Study Guide Copymaster.
  • The activity covers AP Environmental Science primary learning objective ERT-4.C.
  • It will take a total of 2 hours over 3 days to complete this activity (Day 1: 20 minutes, Day 2: 30 minutes, and Day 3: 10 to 15 minutes).

Kit contains:

  • Soil pH Indicator Solution, 50 mL
  • Soil Nutrient Extracting Solution, 500 mL
  • Phosphorous Reagent #2, 20 mL
  • Phosphorous Indicator Powder, 5 g with Scoop
  • Nitrate Reagent #1, 25 mL
  • Nitrate Reagent #2, 5 g with Scoop
  • Potassium Reagent Solution, 30 mL
  • Potassium Indicator Powder, 5 g with Scoop
  • Sand, 3 kg
  • Clay Powder, 3 kg
  • Detergent Powder, 40 g with Spoon
  • 8 Extraction Bottles
  • 8 Jars with Lids
  • 40 Pipettes
  • 11 Soil Scoops (8 for Soil Samples, 3 for Reagents)
  • 24 Coffee Filters; 24 Cups, 5 oz.
  • 8 pH, N, P, K Color Charts

Needed but not supplied:

  • Soil Samples
  • Beakers, 250 mL
  • Test Tubes
  • Graduated Cylinders, 50 to 100 mL
  • Funnels
  • Ring Stand with Metal Rings and Clamp
  • Timer
  • Ruler
  • Markers
  • Sieves
  • Newspaper or Paper Towel for Sieved Soil, optional

Specifications

Grade Level High School
Grade 9 to 12
Class Size 8 Lab Groups
Product Type Properties of Soil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.