Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP repeat Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119810 Supplier Invitrogen™ Supplier No. PA5119810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Recombinant protein lysates.

The C9orf72 'GGGGCC' repeat sequence codes five repeat peptide 'GA repeat; GAGAGAGAGA', 'GP repeat; GPGPGPGPG', 'GR repeat; GRGRGRGRG', 'AP repeat; APAPAPAPA' and 'PR repeat; PRPRPRPRP'. It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP repeat
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias AP repeat; APAPAPAPA; C9RAN; GGGCC; poly AP
Host Species Rabbit
Immunogen Synthetic peptide immune sequence is CAPAPAPAPA.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.